AC5424075961 Accela ChemBio Inc.

2-Benzyl-2,7-diazaspiro[4.5]decane

  • MDL number MFCD11223500
  • Cas Number : 1086395-71-2
  • Molecular Weight : 230.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027020&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027020.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart