AC5424075983 Accela ChemBio Inc.

2-Bromo-4-methyl-5-nitroanisole

  • MDL number MFCD27935373
  • Cas Number : 41447-19-2
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027058&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027058.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart