AC5424076011 Accela ChemBio Inc.

2',6'-Dimethylbiphenyl-3-carbaldehyde

  • MDL number MFCD11895278
  • Cas Number : 691905-26-7
  • Molecular Weight : 210.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027165&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027165.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart