AC5424076017 Accela ChemBio Inc.

5-(Bromomethyl)-2-(methylthio)pyrimidine

  • MDL number MFCD24465277
  • Cas Number : 323591-23-7
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027177&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY027177.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart