AC5424076022 Accela ChemBio Inc.

2-Bromo-4,6-dimethoxybenzaldehyde

  • MDL number MFCD11110636
  • Cas Number : 81574-69-8
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027196&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027196.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart