AC5424076024 Accela ChemBio Inc.

6-(Trifluoromethyl)salicylaldehyde

  • MDL number MFCD16999701
  • Cas Number : 58914-35-5
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027214&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027214.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart