AC5424076026 Accela ChemBio Inc.

4-(1,1-Dioxido-2-isothiazolidinyl)phenylboronic Acid Pinacol Ester

  • MDL number MFCD26937568
  • Cas Number : 756520-72-6
  • Molecular Weight : 323.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027218&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027218.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart