AC5424076039 Accela ChemBio Inc.

4-Acetamido-3-fluorophenylboronic Acid

  • MDL number MFCD26141998
  • Cas Number : 626251-12-5
  • Molecular Weight : 196.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027268&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027268.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart