AC5424076056 Accela ChemBio Inc.

2-(Boc-aminomethyl)benzyl Bromide

  • MDL number MFCD20257492
  • Cas Number : 220364-33-0
  • Molecular Weight : 300.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027368&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027368.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart