AC5424076067 Accela ChemBio Inc.

3-Fluoro-5-methoxybenzylamine

  • MDL number MFCD04116363
  • Cas Number : 914465-85-3
  • Molecular Weight : 155.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027486&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027486.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart