AC5424076157 Accela ChemBio Inc.

Ethyl 2,2-Difluoro-3-(methylamino)propanoate Hydrochloride

  • MDL number MFCD27986811
  • Cas Number : 1956311-12-8
  • Molecular Weight : 203.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028887&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028887.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart