AC5424076178 Accela ChemBio Inc.

6-Bromo-3-isopropyl-1H-indazole

  • MDL number MFCD27987866
  • Cas Number : 1391123-54-8
  • Molecular Weight : 239.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY029530&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY029530.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart