AC5424076187 Accela ChemBio Inc.

4-Chloro-2-methylbutanoyl Chloride

  • MDL number MFCD19232042
  • Cas Number : 51708-72-6
  • Molecular Weight : 155.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027796&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027796.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart