AC5424076188 Accela ChemBio Inc.

4-Chloro-3-methylbutanoyl Chloride

  • MDL number MFCD19232353
  • Cas Number : 63480-13-7
  • Molecular Weight : 155.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027797&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027797.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart