AC5424076191 Accela ChemBio Inc.

5-(4-Hydroxyphenyl)thiophene-2-carboxylic Acid

  • MDL number MFCD27500654
  • Cas Number : 116016-57-0
  • Molecular Weight : 220.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027812&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027812.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart