AC5424076192 Accela ChemBio Inc.

5-Bromo-3-methylthiophene-2-carbaldehyde

  • MDL number MFCD10000783
  • Cas Number : 38239-46-2
  • Molecular Weight : 205.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027813&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027813.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart