AC5424076199 Accela ChemBio Inc.

2-Aminoimidazole-4-carboxylic Acid

  • MDL number MFCD08668249
  • Cas Number : 860011-60-5
  • Molecular Weight : 127.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027829&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027829.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart