AC5424076219 Accela ChemBio Inc.

2-Amino-1,6-dimethylbenzimidazole

  • MDL number MFCD20703217
  • Cas Number : 945023-34-7
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027889&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027889.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart