AC5424076224 Accela ChemBio Inc.

N,N-Dimethylpiperidine-3-carboxamide

  • MDL number MFCD09802939
  • Cas Number : 5505-20-4
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027894&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027894.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart