AC5424076239 Accela ChemBio Inc.

3-(4-Methyl-1-piperazinyl)phenylboronic Acid

  • MDL number MFCD13194680
  • Cas Number : 1139717-76-2
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027977&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027977.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart