AC5424076242 Accela ChemBio Inc.

5-Bromo-1,1,1,2,2-pentafluoropentane

  • MDL number MFCD27935381
  • Cas Number : 1383437-60-2
  • Molecular Weight : 241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027994&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027994.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart