AC5424076251 Accela ChemBio Inc.

4-Amino-2,6-dimethoxypyridine

  • MDL number MFCD19688654
  • Cas Number : 17325-39-2
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028009&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028009.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart