AC5424076256 Accela ChemBio Inc.

2-Bromo-4-(difluoromethoxy)benzonitrile

  • MDL number MFCD18390874
  • Cas Number : 1261818-72-7
  • Molecular Weight : 248.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028039&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028039.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart