AC5424076258 Accela ChemBio Inc.

Ethyl 1-Cyclopropyl-2,4-dioxopiperidine-3-carboxylate

  • MDL number MFCD27935386
  • Cas Number : 1453423-57-8
  • Molecular Weight : 225.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028043&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028043.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart