AC5424076265 Accela ChemBio Inc.

2-Fluoro-1-iodo-2-methylpropane

  • MDL number MFCD24458933
  • Cas Number : 19869-79-5
  • Molecular Weight : 202.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028245&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028245.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart