AC5424076266 Accela ChemBio Inc.

4-Amino-6-chlorocinnoline

  • MDL number MFCD20691253
  • Cas Number : 18259-66-0
  • Molecular Weight : 179.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028257&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY028257.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart