AC5424076271 Accela ChemBio Inc.

1-Hydroxy-1-phenyl-2-propanone

  • MDL number MFCD03792883
  • Cas Number : 90-63-1
  • Molecular Weight : 150.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028287&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028287.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart