AC5424076364 Accela ChemBio Inc.

3-(1-Pyrrolidinyl)aniline

  • MDL number MFCD05259334
  • Cas Number : 115833-93-7
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Colorless to brown liquid, or white to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028684&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P302+P352-P261-P301+P312-P304+P340-P280-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY028684.pdf
hstatements H312-H302-H315-H335-H319-H332
Availability details for
Quantity
  • Availability
  • Add to Cart