AC5424076438 Accela ChemBio Inc.

3-Cyclopropoxy-5-(trifluoromethyl)aniline

  • MDL number MFCD17008595
  • Cas Number : 1243471-01-3
  • Molecular Weight : 217.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028827&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028827.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart