AC5424076441 Accela ChemBio Inc.

3-Acetamido-5-hydroxybenzoic Acid

  • MDL number MFCD20654260
  • Cas Number : 93561-88-7
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028830&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028830.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart