AC5424076458 Accela ChemBio Inc.

3-Phenylpyrazin-2(1H)-one

  • MDL number MFCD24394632
  • Cas Number : 2882-18-0
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027698&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027698.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart