AC5424076469 Accela ChemBio Inc.

5-Fluoro-2-(methylsulfonyl)benzonitrile

  • MDL number MFCD18378491
  • Cas Number : 1379097-18-3
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028941&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028941.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart