AC5424076472 Accela ChemBio Inc.

5-Aminothiazolo[4,5-d]pyrimidin-2(3H)-one

  • MDL number MFCD17017931
  • Cas Number : 848691-22-5
  • Molecular Weight : 168.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028954&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028954.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart