AC5424076478 Accela ChemBio Inc.

2-[2-(Methylsulfonyl)phenyl]ethylamine

  • MDL number MFCD13190319
  • Cas Number : 933705-18-1
  • Molecular Weight : 199.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to brown liquid, or white to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028986&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028986.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart