AC5424076501 Accela ChemBio Inc.

[1,1':4',1''-Terphenyl]-4,4''-dicarboxylic Acid

  • MDL number MFCD00389774
  • Cas Number : 13653-84-4
  • Molecular Weight : 318.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027719&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027719.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart