AC5424076524 Accela ChemBio Inc.

5-Bromo-7-methoxybenzofuran

  • MDL number MFCD11877831
  • Cas Number : 164414-62-4
  • Molecular Weight : 227.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027737&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027737.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart