AC5424076556 Accela ChemBio Inc.

4-Aza-9-fluorenone

  • MDL number MFCD00022194
  • Cas Number : 3882-46-0
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY029462&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY029462.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart