AC5424076588 Accela ChemBio Inc.

5-Amino-6-fluoro-1,3-benzodioxole

  • MDL number MFCD22069751
  • Cas Number : 1366234-03-8
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY029694&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY029694.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart