AC5428452546 Accela ChemBio Inc.

4-Fluorobenzenesulphonic Acid

  • MDL number MFCD06658253
  • Cas Number : 368-88-7
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005790&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY005790.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart