AC5428452553 Accela ChemBio Inc.

4-(Methylsulfonyl)-2-pyrrolidinobenzoic Acid

  • MDL number MFCD13191613
  • Cas Number : 1197193-17-1
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006924&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006924.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart