AC5428452555 Accela ChemBio Inc.

3-Amino-4,6-difluoro-1H-indazole

  • MDL number MFCD13191634
  • Cas Number : 1197193-24-0
  • Molecular Weight : 169.1315064

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY007055&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY007055.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart