AC5428452569 Accela ChemBio Inc.

Ethyl 3-Amino-2,2-difluoropropanoate Hydrochloride

  • MDL number MFCD13185039
  • Cas Number : 1263284-04-3
  • Molecular Weight : 189.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY009755&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY009755.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart