AC5428452571 Accela ChemBio Inc.

2-Amino-5-bromo-4-cyclopropylthiazole

  • MDL number MFCD13193636
  • Cas Number : 869854-12-6
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011064&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY011064.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart