AC5428452576 Accela ChemBio Inc.

3-Fluorophenoxyacetonitrile

  • MDL number MFCD09928980
  • Cas Number : 135290-20-9
  • Molecular Weight : 151.1377432

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012355&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012355.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart