AC5428452577 Accela ChemBio Inc.

4-(1-Methyl-5-imidazolyl)benzoic Acid

  • MDL number MFCD18375275
  • Cas Number : 305806-38-6
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012406&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012406.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart