AC5428452586 Accela ChemBio Inc.

3-Bromo-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD17215706
  • Cas Number : 1378861-55-2
  • Molecular Weight : 202.05182

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014003&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY014003.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart