AC5428452588 Accela ChemBio Inc.

N-Boc-3-bromo-4-methylindole

  • MDL number MFCD18911743
  • Cas Number : 1305320-64-2
  • Molecular Weight : 310.18634

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014193&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014193.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart