AC5428452589 Accela ChemBio Inc.

1-Boc-7-fluoro-1H-indazole

  • MDL number MFCD18911746
  • Cas Number : 1305320-59-5
  • Molecular Weight : 236.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014201&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014201.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart