AC5428452600 Accela ChemBio Inc.

6-[2-(Trifluoromethoxy)phenyl]nicotinic Acid

  • MDL number MFCD06410406
  • Cas Number : 197847-94-2
  • Molecular Weight : 283.2027296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016603&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016603.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart