AC5428452662 Accela ChemBio Inc.

Methyl 1,2,3,6-Tetrahydropyridine-4-carboxylate Hydrochloride

  • MDL number MFCD23703517
  • Cas Number : 70684-82-1
  • Molecular Weight : 177.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023222&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023222.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart